Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001323614.1 | NP_001310543.1 | 7 | 885 | |
NM_001323615.1 | NP_001310544.1 | 7 | 885 | ||
NM_001323616.1 | NP_001310545.1 | 8 | 1017 | ||
NM_001323617.1 | NP_001310546.1 | 8 | 852 | ||
NM_015327.2 | NP_056142.2 | 7 | 824 | ||
XM_017000842.1 | XP_016856331.1 | 3 | 770 | ||
GenBank mRNA | AB029012.2 | - | 7 | 826 | |
AB085691.1 | - | 7 | 752 | ||
AK092183.1 | - | 7 | 800 | ||
AY168922.1 | - | 7 | 678 | ||
AY336728.1 | - | 7 | 675 | ||
BC038296.1 | - | 7 | 825 | 4824487 |