Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
54749
Gene Name
ependymin related 1
Gene Aliases
EPDR, MERP-1, MERP1, UCC1
Chromosome Location
Chr. 7: 37920561 - 37951941 on Build GRCh38
UniGene ID
Hs.563491
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242946.1 NP_001229875.2 1 258
NM_017549.4 NP_060019.2 1 258
GenBank mRNA AY027862.2 - 1 258
BC018299.1 - 1 150 3461888

View Full Product Details