Gene Transcripts
- Gene Symbol
- MBTD1
- Entrez Gene ID
- 54799
- Gene Name
- mbt domain containing 1
- Gene Aliases
- SA49P01
- Chromosome Location
- Chr. 17: 51177419 - 51260511 on Build GRCh38
- UniGene ID
- Hs.656803
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017643.2 |
NP_060113.2
|
7, 8 |
956 |
|
XM_005257467.4 |
XP_005257524.1
|
6, 7 |
901 |
|
XM_005257468.4 |
XP_005257525.1
|
6, 7 |
894 |
|
XM_011524922.2 |
XP_011523224.1
|
6, 7 |
903 |
|
XM_011524923.2 |
XP_011523225.1
|
6, 7 |
900 |
|
XM_011524924.2 |
XP_011523226.1
|
6, 7 |
896 |
|
XM_011524925.1 |
XP_011523227.1
|
7, 8 |
1017 |
|
XM_011524926.2 |
XP_011523228.1
|
7, 8 |
1121 |
|
XM_011524928.2 |
XP_011523230.1
|
6, 7 |
1058 |
|
XM_011524929.2 |
XP_011523231.1
|
7, 8 |
972 |
|
XM_011524930.1 |
XP_011523232.1
|
6, 7 |
694 |
|
XM_011524931.2 |
XP_011523233.1
|
5, 6 |
528 |
|
XM_011524933.2 |
XP_011523235.1
|
8, 9 |
1078 |
|
XM_011524934.2 |
XP_011523236.1
|
6, 7 |
902 |
|
XM_011524935.2 |
XP_011523237.1
|
6, 7 |
895 |
|
XM_011524936.1 |
XP_011523238.1
|
6, 7 |
694 |
|
XM_017024782.1 |
XP_016880271.1
|
7, 8 |
1268 |
|
GenBank mRNA |
AK000062.1 |
- |
3, 4 |
288 |
|
AK124061.1 |
- |
6, 7 |
694 |
|
BC034364.1 |
- |
7, 8 |
927 |
|
BC101736.1 |
- |
2, 3 |
217 |
8069242 |
View Full Product Details