Gene Transcripts

Gene Symbol
Plekhm1
Entrez Gene ID
303584
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 1
Gene Aliases
Chromosome Location
Chr. 10: 91451890 - 91500675 on Build Rnor_6.0
UniGene ID
Rn.101335
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009677.1 NP_001009677.1 9, 10 2937
XM_006247527.2 XP_006247589.1 9, 10 2977
XM_008768292.1 XP_008766514.1 8, 9 2757
GenBank mRNA BC087058.1 - 9, 10 2937

View Full Product Details