Gene Transcripts

Gene Symbol
Timd2
Entrez Gene ID
287222
Gene Name
T-cell immunoglobulin and mucin domain containing 2
Gene Aliases
Tim2
Chromosome Location
Chr. 10: 31755473 - 31796423 on Build Rnor_6.0
UniGene ID
Rn.136665
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013855.1 NP_001013877.1 3 367
XM_006246168.1 XP_006246230.1 4 652
XM_008767667.1 XP_008765889.1 4 415
XM_008767668.1 XP_008765890.1 4 415
XM_008767669.1 XP_008765891.1 4 907
GenBank mRNA BC089832.1 - 3 367

View Full Product Details