Gene Transcripts

Gene Symbol
Pigu
Entrez Gene ID
228812
Gene Name
phosphatidylinositol glycan anchor biosynthesis, class U
Gene Aliases
5430426F17Rik, BC028278, Cdc91l1
Chromosome Location
Chr. 2: 155278243 - 155357424 on Build GRCm38
UniGene ID
Mm.5434
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004721.1 NP_001004721.1 1 105
GenBank mRNA AK140955.1 - 1 101
AK143517.1 - 1 105
AK162913.1 - 80
AK171988.1 - 1 84
BB839941.1 - 1 105
BC120680.1 - 1 94 40129603
BC120682.1 - 1 94 40129605
CK334159.1 - 1 58

View Full Product Details