Gene Transcripts

Gene Symbol
KNOP1
Entrez Gene ID
400506
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
101F10.1, C16orf88, FAM191A, TSG118
Chromosome Location
Chr. 16: 19706347 - 19718240 on Build GRCh38
UniGene ID
Hs.585209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012991.2 NP_001013009.2 4 1156
XM_005255317.4 XP_005255374.1 6 1351
XM_006721046.3 XP_006721109.1 5 1514
XM_011545855.1 XP_011544157.1 5 1400
XM_017023231.1 XP_016878720.1 5 1292
XM_017023232.1 XP_016878721.1 4 1437
GenBank mRNA BC117562.1 - 4 1156

View Full Product Details