Gene Transcripts

Gene Symbol
Bin3
Entrez Gene ID
361065
Gene Name
bridging integrator 3
Gene Aliases
Chromosome Location
Chr. 15: 51799339 - 51818697 on Build Rnor_6.0
UniGene ID
Rn.153573
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013186.1 NP_001013204.1 7 528
GenBank mRNA BC079146.1 - 7 528

View Full Product Details