Gene Transcripts
- Gene Symbol
- OR5AC2
- Entrez Gene ID
- 81050
- Gene Name
- olfactory receptor family 5 subfamily AC member 2
- Gene Aliases
- HSA1
- Chromosome Location
- Chr. 3: 98087173 - 98088102 on Build GRCh38
- UniGene ID
- Hs.554715
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_054106.1 |
NP_473447.1
|
1 |
67 |
|
View Full Product Details