Gene Transcripts

Gene Symbol
Utp3
Entrez Gene ID
305258
Gene Name
UTP3, small subunit (SSU) processome component, homolog (S. cerevisiae)
Gene Aliases
Crlz1
Chromosome Location
Chr. 14: 21153998 - 21155652 on Build Rnor_6.0
UniGene ID
Rn.23516
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012036.1 NP_001012036.1 1 961
GenBank mRNA BC079277.1 - 1 961

View Full Product Details