Gene Transcripts

Gene Symbol
Adck3
Entrez Gene ID
360887
Gene Name
aarF domain containing kinase 3
Gene Aliases
Cabc1
Chromosome Location
Chr. 13: 98451629 - 98480438 on Build Rnor_6.0
UniGene ID
Rn.160865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013185.1 NP_001013203.1 7, 8 985
XM_006250387.1 XP_006250449.1 7, 8 1068
GenBank mRNA BC091388.1 - 7, 8 985

View Full Product Details