Gene Transcripts

Gene Symbol
Podnl1
Entrez Gene ID
244550
Gene Name
podocan-like 1
Gene Aliases
5832418A03Rik, Gm506
Chromosome Location
Chr. 8: 84125927 - 84143016 on Build GRCm38
UniGene ID
Mm.334654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013384.2 NP_001013402.2 9 1621
GenBank mRNA AK077813.1 - 3 2480
BC063764.1 - 9 1621 30288186
DQ303463.1 - 9 1606

View Full Product Details