Gene Transcripts
- Gene Symbol
- ANO1
- Entrez Gene ID
- 55107
- Gene Name
- anoctamin 1
- Gene Aliases
- DOG1, ORAOV2, TAOS2, TMEM16A
- Chromosome Location
- Chr. 11: 70078302 - 70189545 on Build GRCh38
- UniGene ID
- Hs.503074
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018043.5 |
NP_060513.5
|
12, 14 |
1654 |
|
XM_006718602.2 |
XP_006718665.1
|
13, 15 |
1436 |
|
XM_006718604.2 |
XP_006718667.1
|
13, 15 |
1551 |
|
XM_011545121.2 |
XP_011543423.1
|
13, 15 |
1428 |
|
XM_011545125.2 |
XP_011543427.1
|
13, 15 |
1428 |
|
XM_011545127.2 |
XP_011543429.1
|
12, 14 |
1362 |
|
XM_011545129.2 |
XP_011543431.1
|
13, 15 |
1343 |
|
GenBank mRNA |
AB845669.1 |
- |
12, 14 |
1349 |
|
AK001123.1 |
- |
3, 5 |
270 |
|
AK293088.1 |
- |
12, 14 |
1362 |
|
AL833582.1 |
- |
12, 14 |
1398 |
|
AY728143.1 |
- |
12, 14 |
1654 |
|
BC027590.2 |
- |
7, 9 |
606 |
|
View Full Product Details