Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026549.3 | NP_080825.1 | 5 | 749 | |
GenBank mRNA | AK003339.1 | - | 5 | 758 | |
AK077948.1 | - | 5 | 750 | ||
AK151495.1 | - | 5 | 753 | ||
AK154811.1 | - | 5 | 765 | ||
BC025090.1 | - | 2 | 213 | ||
BC038524.1 | - | 2 | 219 | ||
BC115797.1 | - | 5 | 748 | 40103840 |