Gene Transcripts

Gene Symbol
AAED1
Entrez Gene ID
195827
Gene Name
AhpC/TSA antioxidant enzyme domain containing 1
Gene Aliases
C9orf21
Chromosome Location
Chr. 9: 96641251 - 96655331 on Build GRCh38
UniGene ID
Hs.44640
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153698.1 NP_714542.1 3 319
XM_005251783.3 XP_005251840.1 3 314
XM_005251784.4 XP_005251841.1 3 171
GenBank mRNA BC136501.1 - 3 157
BC136503.1 - 3 319 9020492
BF445089.1 - 3 194
BM843088.1 - 3 319

View Full Product Details