Gene Transcripts

Gene Symbol
AGGF1
Entrez Gene ID
55109
Gene Name
angiogenic factor with G-patch and FHA domains 1
Gene Aliases
GPATC7, GPATCH7, HSU84971, HUS84971, VG5Q
Chromosome Location
Chr. 5: 77030385 - 77065234 on Build GRCh38
UniGene ID
Hs.634849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018046.4 NP_060516.2 12 2194
GenBank mRNA AK001145.1 - 7 706
AK289394.1 - 8 1046
AY500994.1 - 12 2113
BC015478.1 - 7 945
BC029382.1 - 5 503
BC032844.1 - 12 2180 5266978
BX648120.1 - 8 1003

View Full Product Details