Gene Transcripts

Gene Symbol
C15orf61
Entrez Gene ID
145853
Gene Name
chromosome 15 open reading frame 61
Gene Aliases
Chromosome Location
Chr. 15: 67521184 - 67527303 on Build GRCh38
UniGene ID
Hs.438385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143936.1 NP_001137408.1 1, 2 415
GenBank mRNA AK294012.1 - 1, 2 350
AY726587.1 - 1, 2 307

View Full Product Details