Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040032.1 | NP_001035121.1 | 4 | 784 | |
XM_011543816.2 | XP_011542118.1 | 4 | 885 | ||
GenBank mRNA | BC065837.1 | - | 4 | 784 | |
BC132852.1 | - | 4 | 784 | 40146874 | |
BC132854.1 | - | 4 | 784 | 40146876 |