Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018317.3 | NP_060787.2 | 4, 5 | 562 | |
XM_011513852.2 | XP_011512154.1 | 4, 5 | 402 | ||
XM_017008363.1 | XP_016863852.1 | 4, 5 | 402 | ||
XM_017008364.1 | XP_016863853.1 | 4, 5 | 402 | ||
GenBank mRNA | AK001944.1 | - | 4, 5 | 402 | |
AK225395.1 | - | 4, 5 | 402 | ||
AK289631.1 | - | 4, 5 | 402 | ||
AK310795.1 | - | 4, 5 | 409 | ||
BC031642.1 | - | 4, 5 | 358 | 5175317 |