Gene Transcripts

Gene Symbol
C4orf47
Entrez Gene ID
441054
Gene Name
chromosome 4 open reading frame 47
Gene Aliases
Chromosome Location
Chr. 4: 185405227 - 185449828 on Build GRCh38
UniGene ID
Hs.549398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114357.1 NP_001107829.1 2 177
XM_005263019.3 XP_005263076.1 4 530
XM_011531989.2 XP_011530291.1 3 456
XM_017008236.1 XP_016863725.1 4 1923
XM_017008237.1 XP_016863726.1 5 698
XM_017008238.1 XP_016863727.1 3 1707
XM_017008239.1 XP_016863728.1 3 1710
XM_017008240.1 XP_016863729.1 3 459
XM_017008242.1 XP_016863731.1 4 1989
GenBank mRNA AY947525.1 - 1 47
BC150574.1 - 2 177 40127870

View Full Product Details