Gene Transcripts

Gene Symbol
C12orf77
Entrez Gene ID
196415
Gene Name
chromosome 12 open reading frame 77
Gene Aliases
Chromosome Location
Chr. 12: 24993424 - 24997673 on Build GRCh38
UniGene ID
Hs.434453
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101339.2 NP_001094809.2 3 539
XM_017018966.1 XP_016874455.1 4 550
GenBank mRNA BC035880.1 - 3 550

View Full Product Details