Gene Transcripts
- Gene Symbol
- CDCA7L
- Entrez Gene ID
- 55536
- Gene Name
- cell division cycle associated 7 like
- Gene Aliases
- JPO2, R1, RAM2
- Chromosome Location
- Chr. 7: 21900899 - 21945924 on Build GRCh38
- UniGene ID
- Hs.744069
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127370.2 |
NP_001120842.1
|
3, 4 |
350 |
|
NM_018719.4 |
NP_061189.2
|
2, 3 |
296 |
|
GenBank mRNA |
AK022955.1 |
- |
2, 3 |
271 |
|
AK095965.1 |
- |
3, 4 |
304 |
|
AK292490.1 |
- |
2, 3 |
271 |
|
AY161168.1 |
- |
2, 3 |
173 |
|
AY161169.1 |
- |
2, 3 |
176 |
|
BC009352.2 |
- |
2, 3 |
188 |
4139111 |
BC014630.2 |
- |
1, 2 |
140 |
|
BC025242.1 |
- |
2, 3 |
250 |
4842169 |
BC032576.1 |
- |
2, 3 |
261 |
5491780 |
BC039823.1 |
- |
2, 3 |
260 |
6057601 |
DA041070.1 |
- |
2, 3 |
296 |
|
View Full Product Details