Gene Transcripts

Gene Symbol
PCDHB4
Entrez Gene ID
56131
Gene Name
protocadherin beta 4
Gene Aliases
PCDH-BETA4
Chromosome Location
Chr. 5: 141121750 - 141125623 on Build GRCh38
UniGene ID
Hs.591250
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018938.3 NP_061761.1 1 404
GenBank mRNA AF152497.1 - 1 155
AF217754.1 - 1 155
AK313640.1 - 1 314
BC098139.1 - 1 155 40008131
BC098346.1 - 1 155 40008130
DA231000.1 - 1 405
DA409037.1 - 1 343

View Full Product Details