Gene Transcripts

Gene Symbol
ZNF469
Entrez Gene ID
84627
Gene Name
zinc finger protein 469
Gene Aliases
BCS, BCS1
Chromosome Location
Chr. 16: 88424805 - 88440757 on Build GRCh38
UniGene ID
Hs.54925
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127464.2 NP_001120936.2 1 2525
XM_017023784.1 XP_016879273.1 2 3675
XM_017023785.1 XP_016879274.1 2 2718

View Full Product Details