Gene Transcripts

Gene Symbol
C1QTNF8
Entrez Gene ID
390664
Gene Name
C1q and tumor necrosis factor related protein 8
Gene Aliases
CTRP8, UNQ5829
Chromosome Location
Chr. 16: 1088226 - 1096306 on Build GRCh38
UniGene ID
Hs.527853
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207419.3 NP_997302.2 5 2572
GenBank mRNA AY358832.1 - 6 2170

View Full Product Details