Gene Transcripts

Gene Symbol
Bloc1s2
Entrez Gene ID
293938
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 2
Gene Aliases
RSEP1, Sep1
Chromosome Location
Chr. 1: 263913901 - 263920980 on Build Rnor_6.0
UniGene ID
Rn.19119
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037349.2 NP_001032426.1 5 512
XM_008760400.1 XP_008758622.1 5 539
XM_008760401.1 XP_008758623.1 6 614
GenBank mRNA AY786315.1 - 5 493
EF061226.2 - 6 568

View Full Product Details