Gene Transcripts

Gene Symbol
Pebp4
Entrez Gene ID
73523
Gene Name
phosphatidylethanolamine binding protein 4
Gene Aliases
PEBP-4
Chromosome Location
Chr. 14: 69840407 - 70059918 on Build GRCm38
UniGene ID
Mm.23509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028526.4 NP_082802.1 3 431
NM_028560.4 NP_082836.2 3 431
GenBank mRNA AK006750.1 - 3 432
AK006964.1 - 3 422
BC048621.1 - 3 409 6742909

View Full Product Details