Gene Transcripts

Gene Symbol
Ugt2b36
Entrez Gene ID
231396
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide B36
Gene Aliases
Chromosome Location
Chr. 5: 87065927 - 87092555 on Build GRCm38
UniGene ID
Mm.439851
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029867.1 NP_001025038.1 1 211
GenBank mRNA AK149425.1 - 1 214
BC141232.1 - 1 206 9055844
BC145555.1 - 1 206 9054119

View Full Product Details