Gene Transcripts

Gene Symbol
Psg26
Entrez Gene ID
574429
Gene Name
pregnancy-specific glycoprotein 26
Gene Aliases
EG574429, cea14
Chromosome Location
Chr. 7: 18474577 - 18484197 on Build GRCm38
UniGene ID
Mm.296754
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029893.1 NP_001025064.1 5 1976
GenBank mRNA BC099404.1 - 5 1976

View Full Product Details