Gene Transcripts

Gene Symbol
MSS51
Entrez Gene ID
118490
Gene Name
MSS51 mitochondrial translational activator
Gene Aliases
ZMYND17
Chromosome Location
Chr. 10: 73423579 - 73433561 on Build GRCh38
UniGene ID
Hs.631823
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024593.1 NP_001019764.1 7 2012
GenBank mRNA AK096884.1 - 7 1582
BC094693.1 - 7 1991 30529259
BX537678.1 - 5 3137
CR936824.1 - 6 2319

View Full Product Details