Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010852.3 | NP_001010852.2 | 3 | 1779 | |
XM_017010266.1 | XP_016865755.1 | 3 | 3481 | ||
GenBank mRNA | AK095527.1 | - | 3 | 1783 | |
AK295158.1 | - | 3 | 241 | ||
BC101376.1 | - | 3 | 1313 | 40025886 | |
BC101377.2 | - | 3 | 1315 | 40025889 |