Gene Transcripts

Gene Symbol
C3orf14
Entrez Gene ID
57415
Gene Name
chromosome 3 open reading frame 14
Gene Aliases
HT021
Chromosome Location
Chr. 3: 62318973 - 62336213 on Build GRCh38
UniGene ID
Hs.47166
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291941.1 NP_001278870.1 4 335
NM_001291942.1 NP_001278871.1 3 223
NM_001291943.1 NP_001278872.1 4 335
NM_020685.4 NP_065736.1 4 442
XM_017006931.1 XP_016862420.1 4 818
GenBank mRNA AF236158.1 - 4 272
AK310431.1 - 4 413
AK313915.1 - 4 442
BC017772.2 - 3 252 4307022
BU568674.1 - 177
DA615061.1 - 4 293
DA783189.1 - 4 335

View Full Product Details