Gene Transcripts

Gene Symbol
Psme2
Entrez Gene ID
19188
Gene Name
proteasome (prosome, macropain) activator subunit 2 (PA28 beta)
Gene Aliases
AA589371, AI788882, PA28b
Chromosome Location
Chr. 14: 55587440 - 55591146 on Build GRCm38
UniGene ID
Mm.371573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029855.1 NP_001025026.1 11 749
NM_011190.3 NP_035320.1 11 849
GenBank mRNA AA216967.1 - 2 127
AK012344.1 - 11 778
AK088654.1 - 11 776
AK143556.1 - 11 781
BC002301.1 - 3 192
BC005680.1 - 11 769 3154255
BC057859.1 - 11 846 5310526

Gene Symbol
Psme2b
Entrez Gene ID
621823
Gene Name
protease (prosome, macropain) activator subunit 2B
Gene Aliases
PA28b-p, PA28b2, Psme2-like, Psme2b-ps
Chromosome Location
Chr. 11: 48945345 - 48946410 on Build GRCm38
UniGene ID
Mm.440151
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001281472.1 NP_001268401.1 1 1009
GenBank mRNA BB844325.1 - 1 36
D87910.1 - 1 736

View Full Product Details