Gene Transcripts

Gene Symbol
Bex2
Entrez Gene ID
12069
Gene Name
brain expressed X-linked 2
Gene Aliases
AL024066, Bex1
Chromosome Location
Chr. X: 136066565 - 136068236 on Build GRCm38
UniGene ID
Mm.94160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009749.2 NP_033879.1 3 402
GenBank mRNA AF097439.1 - 3 276
AK132005.1 - 3 289
BC027529.1 - 3 276 5058536

View Full Product Details