Gene Transcripts

Gene Symbol
BPESC1
Entrez Gene ID
60467
Gene Name
blepharophimosis, epicanthus inversus and ptosis, candidate 1 (non-protein coding)
Gene Aliases
NCRNA00187
Chromosome Location
Chr. 3: 139104185 - 139125167 on Build GRCh38
UniGene ID
Hs.512673
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_026783.3 1 1112
GenBank mRNA AF196865.1 - 1 1111
BC119686.1 - 1 1090
BC119687.1 - 1 1092

View Full Product Details