Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022728.3 | NP_073565.2 | 2 | 439 | |
GenBank mRNA | AF136169.1 | - | 2 | 207 | |
AF248954.1 | - | 2 | 439 | ||
AK055224.1 | - | 1 | 1536 | ||
AK055238.1 | - | 2 | 409 | ||
AK289619.1 | - | 2 | 240 | ||
BC035048.2 | - | 2 | 241 | 5261681 |