Gene Transcripts

Gene Symbol
PODNL1
Entrez Gene ID
79883
Gene Name
podocan like 1
Gene Aliases
SLRR5B
Chromosome Location
Chr. 19: 13931187 - 13953392 on Build GRCh38
UniGene ID
Hs.448497
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146254.1 NP_001139726.1 8 1901
NM_001146255.1 NP_001139727.1 6 1634
NM_024825.3 NP_079101.3 8 1925
XM_017027313.1 XP_016882802.1 7 1782
GenBank mRNA AK027100.1 - 6 1625
AK290731.1 - 8 1858
AK296101.1 - 6 1621
BC057786.1 - 8 1925 30347370

View Full Product Details