Gene Transcripts

Gene Symbol
CCDC170
Entrez Gene ID
80129
Gene Name
coiled-coil domain containing 170
Gene Aliases
C6orf97, bA282P11.1
Chromosome Location
Chr. 6: 151494040 - 151621193 on Build GRCh38
UniGene ID
Hs.660044
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025059.3 NP_079335.2 3 361
XM_011536147.2 XP_011534449.1 3 399
XM_011536148.2 XP_011534450.1 3 402
GenBank mRNA AK026958.1 - 3 274
AK225330.1 - 3 275
BC060803.1 - 3 310 5298914
BI562667.1 - 3 315
DB232703.1 - 3 361

View Full Product Details