Gene Transcripts
- Gene Symbol
- ABHD18
- Entrez Gene ID
- 80167
- Gene Name
- abhydrolase domain containing 18
- Gene Aliases
- C4orf29
- Chromosome Location
- Chr. 4: 127965256 - 128039711 on Build GRCh38
- UniGene ID
- Hs.143995
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK024759.1 |
- |
11 |
1695 |
|
BC034253.1 |
- |
11 |
1761 |
4339231 |
View Full Product Details