Gene Transcripts

Gene Symbol
ACAD10
Entrez Gene ID
80724
Gene Name
acyl-CoA dehydrogenase family member 10
Gene Aliases
Chromosome Location
Chr. 12: 111686053 - 111757107 on Build GRCh38
UniGene ID
Hs.331141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136538.1 NP_001130010.1 15 2435
NM_025247.5 NP_079523.3 14 2342
GenBank mRNA AK092356.1 - 14 2327
AK097425.1 - 8 1292
AL832043.1 - 15 2413
AY323912.1 - 14 2298
BC003698.1 - 10 1499
BC126358.1 - 14 2256 8992074

View Full Product Details