Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001161009.2 | NP_001154481.1 | 4 | 970 | |
NM_001161011.2 | NP_001154483.1 | 4 | 970 | ||
NM_031905.4 | NP_114111.2 | 5 | 1075 | ||
XM_011516601.2 | XP_011514903.1 | 4 | 1148 | ||
XM_011516602.2 | XP_011514904.1 | 4 | 509 | ||
XM_017012684.1 | XP_016868173.1 | 6 | 1256 | ||
XM_017012685.1 | XP_016868174.1 | 6 | 1256 | ||
XM_017012687.1 | XP_016868176.1 | 4 | 488 | ||
GenBank mRNA | AK075500.1 | - | 5 | 816 | |
AK225082.1 | - | 4 | 703 | ||
AK291818.1 | - | 4 | 698 | ||
AK301565.1 | - | 4 | 703 | ||
AY150853.1 | - | 4 | 970 | ||
AY150854.1 | - | 5 | 1075 | ||
BC003586.1 | - | 4 | 690 | 3357276 |