Gene Transcripts

Gene Symbol
SPRTN
Entrez Gene ID
83932
Gene Name
SprT-like N-terminal domain
Gene Aliases
C1orf124, DVC1, PRO4323, spartan
Chromosome Location
Chr. 1: 231337936 - 231355029 on Build GRCh38
UniGene ID
Hs.554892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001010984.3 NP_001010984.1 1 594
NM_001261462.2 NP_001248391.1 1 594
NM_032018.6 NP_114407.3 1 594
XM_006711818.2 XP_006711881.1 1 566
GenBank mRNA AK027317.1 - 1 265
AK027613.1 - 1 274
AL512744.1 - 1 208
AY358611.1 - 1 1427
BC015740.1 - 1 482
BC068478.1 - 1 545 5298048

View Full Product Details