Gene Transcripts

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
Chromosome Location
Chr. 7: 978485 - 1138291 on Build GRCh38
UniGene ID
Hs.653258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134395.1 NP_001127867.1 4, 5 473
NM_001134396.1 NP_001127868.1 4, 5 454
NM_001318252.1 NP_001305181.1 4, 5 1357
NM_032350.5 NP_115726.1 4, 5 483
NR_134537.1 3, 4 459
XM_006715793.2 XP_006715856.1 3, 4 1097
XM_011515580.2 XP_011513882.1 3, 4 1097
XM_011515581.2 XP_011513883.1 4, 5 517
XM_011515582.2 XP_011513884.1 4, 5 468
XM_011515583.2 XP_011513885.1 4, 5 492
XM_011515584.2 XP_011513886.1 4, 5 1173
XM_017012719.1 XP_016868208.1 3, 4 1097
XM_017012720.1 XP_016868209.1 4, 5 477
XM_017012721.1 XP_016868210.1 5, 6 1396
XM_017012722.1 XP_016868211.1 4, 5 516
GenBank mRNA AF495721.1 - 4, 5 1313
BC006224.2 - 4, 5 483 3941780
BF129057.1 - 459
BM762287.1 - 3, 4 403
BM763235.1 - 4, 5 457

View Full Product Details