Gene Transcripts

Gene Symbol
SLC7A3
Entrez Gene ID
84889
Gene Name
solute carrier family 7 member 3
Gene Aliases
ATRC3, CAT-3, CAT3
Chromosome Location
Chr. X: 70925579 - 70931125 on Build GRCh38
UniGene ID
Hs.175220
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048164.2 NP_001041629.1 2 326
NM_032803.5 NP_116192.4 2 350
XM_017029912.1 XP_016885401.1 1 841
GenBank mRNA AF320612.1 - 2 301
AK074865.1 - 2 321
AK075014.1 - 2 326
BC033816.1 - 2 305 5206252

View Full Product Details