Gene Transcripts
- Gene Symbol
- ARHGAP19
- Entrez Gene ID
- 84986
- Gene Name
- Rho GTPase activating protein 19
- Gene Aliases
- Chromosome Location
- Chr. 10: 97222173 - 97292673 on Build GRCh38
- UniGene ID
- Hs.80305
- Species
- Homo sapiens
- Gene Symbol
- ARHGAP19-SLIT1
- Entrez Gene ID
- 100533184
- Gene Name
- ARHGAP19-SLIT1 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 10: 97153042 - 97292673 on Build GRCh38
- UniGene ID
- Hs.80305
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037909.1 |
|
5 |
691 |
|
GenBank mRNA |
AY336750.1 |
- |
5 |
656 |
|
View Full Product Details