Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033197.2 | NP_149974.2 | 4 | 433 | |
GenBank mRNA | AF364078.1 | - | 4 | 358 | |
AK290243.1 | - | 4 | 415 | ||
AK310964.1 | - | 4 | 357 | ||
AK313176.1 | - | 4 | 351 | ||
AY358595.1 | - | 4 | 345 | ||
BC008429.1 | - | 4 | 348 | 4291561 | |
CD696225.1 | - | 4 | 455 |