Gene Transcripts
- Gene Symbol
- PIH1D2
- Entrez Gene ID
- 120379
- Gene Name
- PIH1 domain containing 2
- Gene Aliases
- Chromosome Location
- Chr. 11: 112052161 - 112074288 on Build GRCh38
- UniGene ID
- Hs.420662
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001082619.1 |
NP_001076088.1
|
2, 3 |
401 |
|
NM_138789.3 |
NP_620144.1
|
2, 3 |
401 |
|
XM_011542593.2 |
XP_011540895.2
|
2, 3 |
307 |
|
XM_017017201.1 |
XP_016872690.1
|
2, 3 |
518 |
|
XM_017017202.1 |
XP_016872691.1
|
2, 3 |
518 |
|
XM_017017203.1 |
XP_016872692.1
|
2, 3 |
518 |
|
XM_017017204.1 |
XP_016872693.1
|
2, 3 |
518 |
|
XM_017017205.1 |
XP_016872694.1
|
2, 3 |
518 |
|
GenBank mRNA |
AK300496.1 |
- |
2, 3 |
264 |
|
BC019238.2 |
- |
1, 2 |
210 |
3935322 |
CB159963.1 |
- |
2, 3 |
401 |
|
View Full Product Details