Gene Transcripts

Gene Symbol
ALS2CR12
Entrez Gene ID
130540
Gene Name
amyotrophic lateral sclerosis 2 chromosome region candidate 12
Gene Aliases
Chromosome Location
Chr. 2: 201288271 - 201357398 on Build GRCh38
UniGene ID
Hs.107944
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127391.2 NP_001120863.1 11 1283
NM_001289993.1 NP_001276922.1 11 951
NM_139163.3 NP_631902.2 11 1283
NR_110620.1 10 1375
XM_011510606.2 XP_011508908.1 11 1283
XM_011510610.2 XP_011508912.1 9 674
XM_011510612.2 XP_011508914.1 5 381
XM_017003360.1 XP_016858849.1 10 866
XM_017003361.1 XP_016858850.1 10 866
XM_017003362.1 XP_016858851.1 9 715
GenBank mRNA AB053314.1 - 10 868
AK124115.1 - 10 1321
AK292283.1 - 11 951
BC031603.1 - 11 1263 5168160
BX095441.1 - 1 29

View Full Product Details