Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113178.2 | NP_001106649.1 | 4 | 701 | |
NM_006599.3 | NP_006590.1 | 3 | 647 | ||
NM_138713.3 | NP_619727.2 | 4 | 701 | ||
NM_138714.3 | NP_619728.2 | 5 | 763 | ||
NM_173214.2 | NP_775321.1 | 4 | 709 | ||
NM_173215.2 | NP_775322.1 | 4 | 709 | ||
XM_006721125.3 | XP_006721188.1 | 3 | 1372 | ||
XM_011522817.2 | XP_011521119.1 | 3 | 647 | ||
XM_011522818.2 | XP_011521120.1 | 6 | 836 | ||
XM_011522819.2 | XP_011521121.1 | 6 | 837 | ||
XM_011522820.2 | XP_011521122.1 | 3 | 267 | ||
XM_011522822.2 | XP_011521124.1 | 2 | 215 | ||
XM_017022870.1 | XP_016878359.1 | 3 | 996 | ||
XM_017022871.1 | XP_016878360.1 | 4 | 723 | ||
XM_017022872.1 | XP_016878361.1 | 3 | 996 | ||
XM_017022873.1 | XP_016878362.1 | 3 | 608 | ||
GenBank mRNA | AB020634.1 | - | 3 | 629 | |
AF089824.1 | - | 4 | 651 | ||
AF134870.1 | - | 1 | 83 | ||
AF163836.1 | - | 2 | 170 | ||
AF346509.1 | - | 5 | 1649 | ||
AJ005683.1 | - | 4 | 325 | ||
AJ005684.1 | - | 4 | 325 | ||
AJ243298.1 | - | 4 | 468 | ||
AJ243299.1 | - | 4 | 460 | ||
BC131509.1 | - | 4 | 605 | 40083218 | |
BC146765.1 | - | 3 | 629 | 9007208 |