Gene Transcripts

Gene Symbol
PRR35
Entrez Gene ID
146325
Gene Name
proline rich 35
Gene Aliases
C16orf11, LA16c-366D1.2
Chromosome Location
Chr. 16: 559786 - 565529 on Build GRCh38
UniGene ID
Hs.632220
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145270.2 NP_660313.1 3 2171
XM_017022959.1 XP_016878448.1 3 2381
XM_017022960.1 XP_016878449.1 4 2904
XM_017022961.1 XP_016878450.1 4 1414
GenBank mRNA BC016613.1 - 3 2171

View Full Product Details